missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichloro-3-ethyl-6-nitrophenol, 98%, Thermo Scientific™

Chemical Identifiers

CAS 99817-36-4
Molecular Formula C8H6Cl2NO3
Molecular Weight (g/mol) 235.04
MDL Number MFCD00270764
InChI Key YTVCECQSAPGJBB-UHFFFAOYSA-M
Synonym 2,4-bis chloranyl-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitrochloride, 2,4-dichloro-3-ethyl-6-nitrophenol, 2-nitro-4,6-dichloro-5-ethylphenol, acmc-209se5, ksc486m4b, phenol, 2,4-dichloro-3-ethyl-6-nitro, pubchem20442
PubChem CID 7020339
IUPAC Name 2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate
SMILES CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC450760050
SDS View Documents Promotion Details
Thermo Fisher Scientific
450760050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

99817-36-4
235.04
YTVCECQSAPGJBB-UHFFFAOYSA-M
7020339
CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O
C8H6Cl2NO3
MFCD00270764
2,4-bis chloranyl-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitrochloride, 2,4-dichloro-3-ethyl-6-nitrophenol, 2-nitro-4,6-dichloro-5-ethylphenol, acmc-209se5, ksc486m4b, phenol, 2,4-dichloro-3-ethyl-6-nitro, pubchem20442
2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate

Specifications

99817-36-4
97.5% min. (HPLC)
MFCD00270764
5g
Solubility in water: insoluble. Other solubilities: soluble in methanol
CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O
235.04
236.05
2, 4-Dichloro-3-ethyl-6-nitrophenol
49.0°C to 51.0°C
C8H6Cl2NO3
Glass bottle
2,4-bis chloranyl-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitrochloride, 2,4-dichloro-3-ethyl-6-nitrophenol, 2-nitro-4,6-dichloro-5-ethylphenol, acmc-209se5, ksc486m4b, phenol, 2,4-dichloro-3-ethyl-6-nitro, pubchem20442
YTVCECQSAPGJBB-UHFFFAOYSA-M
2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate
7020339
98%
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Danger

SDS
Product Certifications

RUO – Research Use Only