missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichloro-3-ethyl-6-nitrophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450760010

 View more versions of this product

Catalog No. AC450760010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 4-Dichloro-3-ethyl-6-nitrophenol
49.0°C to 51.0°C
99817-36-4
C8H7Cl2NO3
Solubility in water: insoluble. Other solubilities: soluble in methanol
CCC1=C(C=C(C(=C1Cl)O)[N+](=O)[O-])Cl
7020339
98%
Glass bottle
1g
97.5% min. (HPLC)
2,4-bis chloranyl-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitrochloride, 2,4-dichloro-3-ethyl-6-nitrophenol, 2-nitro-4,6-dichloro-5-ethylphenol, acmc-209se5, ksc486m4b, phenol, 2,4-dichloro-3-ethyl-6-nitro, pubchem20442
YTVCECQSAPGJBB-UHFFFAOYSA-N
236.05
236.05

Chemical Identifiers

99817-36-4
236.05
2,4-bis chloranyl-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitrochloride, 2,4-dichloro-3-ethyl-6-nitrophenol, 2-nitro-4,6-dichloro-5-ethylphenol, acmc-209se5, ksc486m4b, phenol, 2,4-dichloro-3-ethyl-6-nitro, pubchem20442
CCC1=C(C=C(C(=C1Cl)O)[N+](=O)[O-])Cl
C8H7Cl2NO3
YTVCECQSAPGJBB-UHFFFAOYSA-N
7020339
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Danger

Documents