missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dichloro-3-ethyl-6-nitrophenol, 98%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC450760050
Specifications
2, 4-Dichloro-3-ethyl-6-nitrophenol | |
49.0°C to 51.0°C | |
C8H6Cl2NO3 | |
Glass bottle | |
5g | |
YTVCECQSAPGJBB-UHFFFAOYSA-M | |
2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate | |
7020339 | |
98% |
99817-36-4 | |
97.5% min. (HPLC) | |
MFCD00270764 | |
2,4-bis chloranyl-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitrochloride, 2,4-dichloro-3-ethyl-6-nitrophenol, 2-nitro-4,6-dichloro-5-ethylphenol, acmc-209se5, ksc486m4b, phenol, 2,4-dichloro-3-ethyl-6-nitro, pubchem20442 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol | |
CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O | |
235.04 | |
236.05 |
Chemical Identifiers
99817-36-4 | |
235.04 | |
YTVCECQSAPGJBB-UHFFFAOYSA-M | |
7020339 | |
CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O |
C8H6Cl2NO3 | |
MFCD00270764 | |
2,4-bis chloranyl-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, 2,4-dichloro-3-ethyl-6-nitrochloride, 2,4-dichloro-3-ethyl-6-nitrophenol, 2-nitro-4,6-dichloro-5-ethylphenol, acmc-209se5, ksc486m4b, phenol, 2,4-dichloro-3-ethyl-6-nitro, pubchem20442 | |
2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
GHS Signal Word: Danger
RUO – Research Use Only