Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-3-ethyl-6-nitrophenol, 98%

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-3-ethyl-6-nitrophenol
CAS 99817-36-4
Molecular Formula C8H7Cl2NO3
Formula Weight 236.05
MDL Number MFCD00270764
UN Number UN2811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2226109

 
alfa aesar™
B2226109
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2226188

 
alfa aesar™
B2226188
2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.05
Chemical Name or Material 2,4-Dichloro-3-ethyl-6-nitrophenol
TSCA TSCA
Assay 98%
Molecular Formula C8H7Cl2NO3
MDL Number MFCD00270764
CAS 99817-36-4
UN Number UN2811
Melting Point 49° to 51°C
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P280-P273-P305+P351+P338-P309-P310-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P280-P273-P305+P351+P338-P309-P310-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.