missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-3-cyanopyridine, 95%

Manufacturer:  Alfa Aesar™ H6334703

 View more versions of this product

Catalog No. AAH6334703



Description

Description

Applications
2,4-Dichloro-3-cyanopyridine, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2,4-Dichloro-3-cyanopyridine
180995-12-4
C6H2Cl2N2
UN3439
C1=CN=C(C(=C1Cl)C#N)Cl
172.996
173
1g
95%
MFCD09750302
OQYZEFBWEXEOPL-UHFFFAOYSA-N
2,4-dichloropyridine-3-carbonitrile
21997828

Chemical Identifiers

180995-12-4
172.996
OQYZEFBWEXEOPL-UHFFFAOYSA-N
2,4-dichloropyridine-3-carbonitrile
C6H2Cl2N2
MFCD09750302
21997828
C1=CN=C(C(=C1Cl)C#N)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

SDS Documents