Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-3,5-dinitrobenzotrifluoride, 97%

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-3,5-dinitrobenzotrifluoride
CAS 29091-09-6
Molecular Formula C7HCl2F3N2O4
Formula Weight 305
MDL Number MFCD00042480
UN Number UN3077
Beilstein 2062037
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1410503

  SDS
alfa aesar™
L1410503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1410506

  SDS
alfa aesar™
L1410506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 305
Assay 97%
CAS 29091-09-6
Health Hazard 1 Warning
Chemical Name or Material 2,4-Dichloro-3,5-dinitrobenzotrifluoride
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00042480
Boiling Point 291° to 294°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Molecular Formula C7HCl2F3N2O4
Quantity 1g
UN Number UN3077
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Melting Point 74° to 78°C
TSCA TSCA
Beilstein 2062037
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.