Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-3,5-dimethylphenol, 96%

$188.35 - $625.31

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-3,5-dimethylphenol
CAS 133-53-9
Molecular Formula C8H8Cl2O
Formula Weight 191.06
MDL Number MFCD00019981
Beilstein 2209273
Merck Index 14,3076
Synonym 2,4-Dichloro-3,5-xylenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA173050B

  SDS
alfa aesar™
A173050B
1000g Each for $625.31

AAA1730530

  SDS
alfa aesar™
A1730530
250g Each for $188.35
Description & Specifications

Specifications

Formula Weight 191.06
Beilstein 2209273
MDL Number MFCD00019981
Chemical Name or Material 2,4-Dichloro-3,5-dimethylphenol
Melting Point 95° to 96°C
Assay 96%
Molecular Formula C8H8Cl2O
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 133-53-9
Flash Point 138°C (280°F)
Merck Index 14,3076
Synonym 2,4-Dichloro-3,5-xylenol
TSCA TSCA
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Quantity 1000g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.