Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-1-nitrobenzene, 97%

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-1-nitrobenzene
CAS 611-06-3
Molecular Formula C6H3Cl2NO2
Formula Weight 192.00g/mol
MDL Number MFCD00007071
UN Number UN2811
Beilstein 1451655
Synonym 4-Nitro-m-dichlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB237490B

 
alfa aesar™
B23749-0B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB237490I

 
alfa aesar™
B23749-0I
5000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2374930

 
alfa aesar™
B23749-30
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 258° to 259°C
Melting Point 29° to 33°C
Flash Point 152°C (306°F)
Quantity 1000g
Chemical Name or Material 2,4-Dichloro-1-nitrobenzene
CAS 611-06-3
Assay 97%
Molecular Formula C6H3Cl2NO2
Formula Weight 192.00g/mol
MDL Number MFCD00007071
UN Number UN2811
Beilstein 1451655
Synonym 4-Nitro-m-dichlorobenzene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H302-H317
Toxic in contact with skin.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P280h-P262-P273-P309-P310
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number CZ5420000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P280h-P262-P273-P309-P310
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H317
Toxic in contact with skin.
Harmful if swallowed.
May cause an allergic skin reaction.