Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dichloro-1-iodobenzene, 98%

$54.78 - $138.85

Chemical Identifiers

Chemical Name or Material 2,4-Dichloro-1-iodobenzene
CAS 29898-32-6
Molecular Formula C6H3Cl2I
Formula Weight 272.9
MDL Number MFCD00001035
Beilstein 2245130
Synonym 1,3-Dichloro-4-iodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2068314

 
alfa aesar™
B2068314
25g Each for $54.78

AAB2068322

 
alfa aesar™
B2068322
100g Each for $138.85
Description & Specifications

Specifications

Formula Weight 272.9
Chemical Name or Material 2,4-Dichloro-1-iodobenzene
Assay 98%
Molecular Formula C6H3Cl2I
MDL Number MFCD00001035
CAS 29898-32-6
Boiling Point 262° to 263°C
Beilstein 2245130
Flash Point >110°C (230°F)
Synonym 1,3-Dichloro-4-iodobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.