Please login to your online account to display your discounted pricing

2,4-Dichloro-1-iodobenzene, 97%, Maybridge


Quantity 50g

Manufacturer: fisher scientific company  TL00185EE

 View more versions of this product

Catalog No. TL00185EE

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 272.9
Quantity 50g
Assay 97%
Density 2.0100g/mL
CAS Min % 97.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H3Cl2I
CAS 29898-32-6
Chemical Name or Material 2, 4-Dichloro-1-iodobenzene
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Assay Percent Range 97%

Specifications

Chemical Name or Material 2, 4-Dichloro-1-iodobenzene
CAS 29898-32-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3Cl2I
Formula Weight 272.9
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents