missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichloro-1-(4-nitrophenoxy)benzene, 97%, Maybridge™

$220.09 - $220.09

Chemical Identifiers

CAS 1836-75-5
Molecular Formula C12H7Cl2NO3
Molecular Weight (g/mol) 284.092
MDL Number MFCD00128026
InChI Key XITQUSLLOSKDTB-UHFFFAOYSA-N
Synonym 2,4-dichloro-1-4-nitrophenoxy benzene, mezotox, niclofen, nitrafen, nitraphen, nitrochlor, nitrofen, nitrophen, nitrophene, trizilin
PubChem CID 15787
IUPAC Name 2,4-dichloro-1-(4-nitrophenoxy)benzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
RDP00053EB
SDS View Documents
Fisher Scientific Company
RDP00053EB
25g Amber Glass Bottle
Each for $220.09
 
Description

Description

Specifications

Chemical Identifiers

1836-75-5
284.092
XITQUSLLOSKDTB-UHFFFAOYSA-N
15787
C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl
C12H7Cl2NO3
MFCD00128026
2,4-dichloro-1-4-nitrophenoxy benzene, mezotox, niclofen, nitrafen, nitraphen, nitrochlor, nitrofen, nitrophen, nitrophene, trizilin
2,4-dichloro-1-(4-nitrophenoxy)benzene

Specifications

1.3000g/mL
25g
97.0
C12H7Cl2NO3
MFCD00128026
XITQUSLLOSKDTB-UHFFFAOYSA-N
2,4-dichloro-1-(4-nitrophenoxy)benzene
15787
97%
Amber Glass Bottle
1836-75-5
97%
Cl2C6H3OC6H4NO2
2,4-dichloro-1-4-nitrophenoxy benzene, mezotox, niclofen, nitrafen, nitraphen, nitrochlor, nitrofen, nitrophen, nitrophene, trizilin
C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl
284.092
284.1
2, 4-Dichloro-1-(4-nitrophenoxy)benzene
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
May cause cancer.
May damage the unborn child.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Product Certifications