Please login to your online account to display your discounted pricing

2,4-Dichloro-1-(4-nitrophenoxy)benzene, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RDP00053DA

 View more versions of this product

Catalog No. RDP00053DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 284.1
Quantity 1g
Assay 97%
Density 1.3000g/mL
CAS Min % 97.0
Linear Formula Cl2C6H3OC6H4NO2
Health Hazard 1 GHS Signal Word: Danger
Molecular Formula C12H7Cl2NO3
CAS 1836-75-5
MDL Number MFCD00128026
Chemical Name or Material 2, 4-Dichloro-1-(4-nitrophenoxy)benzene
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
May cause cancer.
May damage the unborn child.

Specifications

Chemical Name or Material 2, 4-Dichloro-1-(4-nitrophenoxy)benzene
CAS 1836-75-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H7Cl2NO3
Linear Formula Cl2C6H3OC6H4NO2
Formula Weight 284.1
MDL Number MFCD00128026
Accessories

GHS Signal Word: Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
May cause cancer.
May damage the unborn child.

SDS & Safety Documents