Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromotoluene, 98%

Chemical Identifiers

Chemical Name or Material 2,4-Dibromotoluene
CAS 31543-75-6
Formula Weight 249.93
MDL Number MFCD00052985
Beilstein 2078741
Synonym 2,4-Dibromo-1-methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6386903

 
alfa aesar™
H6386903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6386906

 
alfa aesar™
H6386906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6386914

 
alfa aesar™
H6386914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.93
Chemical Name or Material 2,4-Dibromotoluene
TSCA TSCA
Refractive Index 1.601
Physical Form Liquid
Assay 98%
MDL Number MFCD00052985
CAS 31543-75-6
Beilstein 2078741
Density 1.85g/mL
Synonym 2,4-Dibromo-1-methylbenzene
Melting Point −10°C (dec.)
Boiling Point 243°C (dec.)
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C