missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromotoluene, 98%

Manufacturer:  Alfa Aesar™ H6386914

 View more versions of this product

Catalog No. AAH6386914



Specifications

Specifications

2,4-Dibromotoluene
1.601
-10°C (decomposition)
31543-75-6
MFCD00052985
2,4-dibromotoluene, benzene, 2,4-dibromo-1-methyl, 2,4-bromotoluene, 2,4-dibromo-toluene, pubchem3915, 2,4-dibromo toluene, acmc-1bn9b, ksc497e7f, 2,4-dibromo-1-methyl-benzene, ghwynnfpugeyem-uhfffaoysa
CC1=C(C=C(C=C1)Br)Br
249.933
249.93
98%
1.85g/mL
243°C (decomposition)
25g
C7H6Br2
2078741
GHWYNNFPUGEYEM-UHFFFAOYSA-N
2,4-dibromo-1-methylbenzene
94421
Liquid

Chemical Identifiers

31543-75-6
249.933
GHWYNNFPUGEYEM-UHFFFAOYSA-N
94421
CC1=C(C=C(C=C1)Br)Br
C7H6Br2
MFCD00052985
2,4-dibromotoluene, benzene, 2,4-dibromo-1-methyl, 2,4-bromotoluene, 2,4-dibromo-toluene, pubchem3915, 2,4-dibromo toluene, acmc-1bn9b, ksc497e7f, 2,4-dibromo-1-methyl-benzene, ghwynnfpugeyem-uhfffaoysa
2,4-dibromo-1-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

storageNote1 : 2° to 8°C

SDS Documents