missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dibromothiophene, 97%, Thermo Scientific™

$30.03 - $30.03

Chemical Identifiers

CAS 3140-92-9
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.928
MDL Number MFCD00043889
InChI Key WAQFYSJKIRRXLP-UHFFFAOYSA-N
Synonym 2,4 dibromothiophene, 2,4-dibromo-thiophene, acmc-1cq64, pubchem1908, thiophene, 2,4-dibromo
PubChem CID 2724560
IUPAC Name 2,4-dibromothiophene
SMILES C1=C(SC=C1Br)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC28510DA
SDS View Documents Promotion Details
Fisher Scientific Company
CC28510DA
1g
Each for $30.03
 
Description

Description

Specifications

Chemical Identifiers

3140-92-9
241.928
WAQFYSJKIRRXLP-UHFFFAOYSA-N
2724560
C1=C(SC=C1Br)Br
C4H2Br2S
MFCD00043889
2,4 dibromothiophene, 2,4-dibromo-thiophene, acmc-1cq64, pubchem1908, thiophene, 2,4-dibromo
2,4-dibromothiophene

Specifications

3140-92-9
100.0
95%
MFCD00043889
1g
WAQFYSJKIRRXLP-UHFFFAOYSA-N
2,4-dibromothiophene
2724560
95%
94.5
2.1400g/mL
C4H2Br2S
Amber Glass Bottle
2,4 dibromothiophene, 2,4-dibromo-thiophene, acmc-1cq64, pubchem1908, thiophene, 2,4-dibromo
C1=C(SC=C1Br)Br
241.928
241.93
2, 4-Dibromothiophene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Product Certifications