Learn More
2,4-Dibromothiophene, 90+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1359106
Description
2,4-Dibromothiophene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Dibromothiophene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.
Solubility
It is insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
2,4-Dibromothiophene | |
2.16 | |
>110°C (230°F) | |
1.632 | |
5 g | |
107651 | |
It is insoluble in water. | |
C1=C(SC=C1Br)Br | |
241.928 | |
241.94 |
3140-92-9 | |
215°C | |
C4H2Br2S | |
MFCD00043889 | |
UN2810 | |
thiophene, 2,4-dibromo, 2,4 dibromothiophene, pubchem1908, 2,4-dibromo-thiophene, acmc-1cq64 | |
WAQFYSJKIRRXLP-UHFFFAOYSA-N | |
2,4-dibromothiophene | |
2724560 | |
≥90% |
Chemical Identifiers
3140-92-9 | |
241.928 | |
WAQFYSJKIRRXLP-UHFFFAOYSA-N | |
2724560 | |
C1=C(SC=C1Br)Br |
C4H2Br2S | |
MFCD00043889 | |
thiophene, 2,4-dibromo, 2,4 dibromothiophene, pubchem1908, 2,4-dibromo-thiophene, acmc-1cq64 | |
2,4-dibromothiophene |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H301-H312+H332-H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only