missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromothiophene, 90+%

Manufacturer:  Alfa Aesar™ L1359114

 View more versions of this product

Catalog No. AAL1359114



Description

Description

Applications
2,4-Dibromothiophene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an intermediate in organic synthesis.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

2,4-Dibromothiophene
215°C
3140-92-9
MFCD00043889
107651
WAQFYSJKIRRXLP-UHFFFAOYSA-N
2,4-dibromothiophene
2724560
≥90%
>110°C (230°F)
25g
C4H2Br2S
UN2810
thiophene, 2,4-dibromo, 2,4 dibromothiophene, pubchem1908, 2,4-dibromo-thiophene, acmc-1cq64
C1=C(SC=C1Br)Br
241.928
241.94

Chemical Identifiers

3140-92-9
241.928
WAQFYSJKIRRXLP-UHFFFAOYSA-N
2724560
C1=C(SC=C1Br)Br
C4H2Br2S
MFCD00043889
thiophene, 2,4-dibromo, 2,4 dibromothiophene, pubchem1908, 2,4-dibromo-thiophene, acmc-1cq64
2,4-dibromothiophene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

Documents