Please login to your online account to display your discounted pricing

2,4-Dibromothiazole, 97+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Dibromothiazole
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C3HBr2NS
Formula Weight 242.91
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392400050

  SDS
thermo fisher scientific
AC392400050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.0%
Physical Form Crystalline Powder
Formula Weight 242.91
Quantity 5g
Color Off-white
Melting Point 78° to 81°C
Infrared Spectrum Authentic
Assay Percent Range 97+%
CAS 4175-77-3
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material 2, 4-Dibromothiazole
Packaging Glass bottle
Molecular Formula C3HBr2NS
Health Hazard 2 GHS P Statement Causes serious eye damage. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation.
Safety and Handling

Warning

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician.

GHS P Statement Causes serious eye damage. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation.