Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromopyrimidine, 95%

$1,041.18 - $1,041.18

Chemical Identifiers

Chemical Name or Material 2,4-Dibromopyrimidine
CAS 3921-01-5
Molecular Formula C4H2Br2N2
Formula Weight 237.88
MDL Number MFCD06798232
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2743306

  SDS
alfa aesar™
H2743306
5g Each for $1,041.18
Description & Specifications

Specifications

Formula Weight 237.88
MDL Number MFCD06798232
CAS 3921-01-5
Assay 95%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Molecular Formula C4H2Br2N2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Chemical Name or Material 2,4-Dibromopyrimidine
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.