missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromophenyl isocyanate, 98%

$87.59 - $266.78

Chemical Identifiers

CAS 55076-90-9
Molecular Formula C7H3Br2NO
Molecular Weight (g/mol) 276.915
MDL Number MFCD00037026
InChI Key CGEZHWIPFKRZBF-UHFFFAOYSA-N
Synonym 2,4-dibromophenyl isocyanate, benzene, 2,4-dibromo-1-isocyanato, acmc-1au9d, 2,4-dibromobenzenisocyanate, 2,4-dibromophenylisocyanate, 2,4-dibromo phenyl isocyanate, 2,4-bis bromanyl-1-isocyanato-benzene
PubChem CID 4649127
IUPAC Name 2,4-dibromo-1-isocyanatobenzene
SMILES C1=CC(=C(C=C1Br)Br)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1871303 View Documents Alfa Aesar™
L1871303
1g Each for $87.59
 
AAL1871306 View Documents Alfa Aesar™
L1871306
5g Each for $266.78
 
Specifications

Chemical Identifiers

55076-90-9
276.915
CGEZHWIPFKRZBF-UHFFFAOYSA-N
4649127
C1=CC(=C(C=C1Br)Br)N=C=O
C7H3Br2NO
MFCD00037026
2,4-dibromophenyl isocyanate, benzene, 2,4-dibromo-1-isocyanato, acmc-1au9d, 2,4-dibromobenzenisocyanate, 2,4-dibromophenylisocyanate, 2,4-dibromo phenyl isocyanate, 2,4-bis bromanyl-1-isocyanato-benzene
2,4-dibromo-1-isocyanatobenzene

Specifications

81°C to 85°C
55076-90-9
MFCD00037026
6501599
CGEZHWIPFKRZBF-UHFFFAOYSA-N
2,4-dibromo-1-isocyanatobenzene
4649127
98%
1g
C7H3Br2NO
UN2811
2,4-dibromophenyl isocyanate, benzene, 2,4-dibromo-1-isocyanato, acmc-1au9d, 2,4-dibromobenzenisocyanate, 2,4-dibromophenylisocyanate, 2,4-dibromo phenyl isocyanate, 2,4-bis bromanyl-1-isocyanato-benzene
C1=CC(=C(C=C1Br)Br)N=C=O
276.915
276.92
2,4-Dibromophenyl isocyanate
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.