Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromophenoxyacetic acid, 97%

$116.07 - $472.55

Chemical Identifiers

Chemical Name or Material 2,4-Dibromophenoxyacetic acid
CAS 10129-78-9
Molecular Formula C8H6Br2O3
Formula Weight 309.95
MDL Number MFCD00051478
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1386506

 
alfa aesar™
A1386506
5g Each for $116.07

AAA1386514

 
alfa aesar™
A1386514
25g Each for $472.55
Description & Specifications

Specifications

Formula Weight 309.95
Chemical Name or Material 2,4-Dibromophenoxyacetic acid
Melting Point 150° to 152°C
Assay 97%
Quantity 5g
Molecular Formula C8H6Br2O3
MDL Number MFCD00051478
CAS 10129-78-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.