missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dibromophenol 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 406511000

 View more versions of this product

Catalog No. AC406511000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 4-Dibromophenol
36° to 39°C
99%
Authentic
100.0
Crystalline Mass, Powder, Crystals and/or Chunks
99%
Br2C6H3OH
MFCD00002149
White to slightly yellow or beige
100g
615-58-7
98.5
Glass bottle
≥98.5%
C6H4Br2O
251.91

Chemical Identifiers

615-58-7
100.0
C6H4Br2O
251.91
2, 4-Dibromophenol, 99%
98.5
99%
Br2C6H3OH
MFCD00002149
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 210-436-5

RTECSNumber : SK8010000

TSCA : TSCA