Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromomesitylene, 98%

$176.30 - $176.30

Chemical Identifiers

Chemical Name or Material 2,4-Dibromomesitylene
CAS 6942-99-0
Molecular Formula C9H10Br2
Formula Weight 278
MDL Number MFCD00013525
Beilstein 2208246
Synonym 2,4-Dibromo-1,3,5-trimethylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1819722

  SDS
alfa aesar™
A1819722
100g Each for $176.30

AAA1819714

  SDS
alfa aesar™
A1819714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2,4-Dibromomesitylene
Boiling Point 278° to 279°C
Melting Point 58° to 62°C
Quantity 100g
CAS 6942-99-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C9H10Br2
Formula Weight 278
MDL Number MFCD00013525
Beilstein 2208246
Synonym 2,4-Dibromo-1,3,5-trimethylbenzene
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.