missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dibromobenzyl alcohol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6474106
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is an halogenated compound for proteomics research. Also used as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
2,4-Dibromobenzyl alcohol | |
C7H6Br2O | |
5 g | |
2,4-dibromophenyl methanol, 2,4-dibromobenzyl alcohol, benzenemethanol, 2,4-dibromo, 2,4-dibromobenzenemethanol, benzenemethanol,2,4-dibromo, pubchem9781, 2,4-dibromobenzylalcohol, acmc-1bfu5, 2,4-bis bromanyl phenyl methanol | |
VXEVXBMLHQPKGA-UHFFFAOYSA-N | |
(2,4-dibromophenyl)methanol | |
17836195 | |
98% |
666747-06-4 | |
MFCD09996906 | |
3241294 | |
Slightly soluble in water. | |
OCC1=C(Br)C=C(Br)C=C1 | |
265.93 | |
265.98 | |
Solid |
Chemical Identifiers
666747-06-4 | |
265.93 | |
VXEVXBMLHQPKGA-UHFFFAOYSA-N | |
(2,4-dibromophenyl)methanol |
C7H6Br2O | |
MFCD09996906 | |
17836195 | |
OCC1=C(Br)C=C(Br)C=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures