missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dibromoanisole, 98%, Thermo Scientific™
$56.80 - $56.80
Chemical Identifiers
CAS | 21702-84-1 |
---|---|
Molecular Formula | C7H6Br2O |
Molecular Weight (g/mol) | 265.92 |
MDL Number | MFCD00000079 |
InChI Key | XGXUGXPKRBQINS-UHFFFAOYSA-N |
Synonym | #, 2,4-dibromo-1-methoxy-benzene, 2,4-dibromoanisole, acmc-209fnf, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, chemwish ic06728, maybridge1_003667, pubchem3847, xgxugxpkrbqins-uhfffaoysa |
PubChem CID | 27011 |
IUPAC Name | 2,4-dibromo-1-methoxybenzene |
SMILES | COC1=C(C=C(C=C1)Br)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC243230500
|
Thermo Fisher Scientific
243230500 |
50g | Glass bottle |
Each for $56.80
|
|
Chemical Identifiers
21702-84-1 | |
265.92 | |
XGXUGXPKRBQINS-UHFFFAOYSA-N | |
27011 | |
COC1=C(C=C(C=C1)Br)Br |
C7H6Br2O | |
MFCD00000079 | |
#, 2,4-dibromo-1-methoxy-benzene, 2,4-dibromoanisole, acmc-209fnf, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, chemwish ic06728, maybridge1_003667, pubchem3847, xgxugxpkrbqins-uhfffaoysa | |
2,4-dibromo-1-methoxybenzene |
Specifications
21702-84-1 | |
100.0 | |
60.0°C to 64.0°C | |
98% | |
C7H6Br2O | |
MFCD00000079 | |
#, 2,4-dibromo-1-methoxy-benzene, 2,4-dibromoanisole, acmc-209fnf, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, chemwish ic06728, maybridge1_003667, pubchem3847, xgxugxpkrbqins-uhfffaoysa | |
XGXUGXPKRBQINS-UHFFFAOYSA-N | |
2,4-dibromo-1-methoxybenzene | |
27011 | |
Crystalline Powder | |
2, 4-Dibromoanisole, 98% |
97.5 | |
Beige | |
101.0°C to 106.0°C (1.0 mmHg) | |
Authentic | |
Br2(C6H3)OCH3 | |
Glass bottle | |
50g | |
COC1=C(C=C(C=C1)Br)Br | |
265.92 | |
265.92 | |
98% |
Safety and Handling
EINECSNumber : 244-536-5
TSCA : TSCA
RUO – Research Use Only