missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dibromoanisole, 98%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific 243230050
Specifications
2, 4-Dibromoanisole | |
21702-84-1 | |
100.0 | |
60.0°C to 64.0°C | |
98% | |
C7H6Br2O | |
MFCD00000079 | |
#, 2,4-dibromo-1-methoxy-benzene, 2,4-dibromoanisole, acmc-209fnf, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, chemwish ic06728, maybridge1_003667, pubchem3847, xgxugxpkrbqins-uhfffaoysa | |
XGXUGXPKRBQINS-UHFFFAOYSA-N | |
2,4-dibromo-1-methoxybenzene | |
27011 | |
Crystalline Powder |
98% | |
97.5 | |
Beige | |
101.0°C to 106.0°C (1.0 mmHg) | |
Authentic | |
Br2(C6H3)OCH3 | |
Glass bottle | |
5g | |
COC1=C(C=C(C=C1)Br)Br | |
265.92 | |
265.92 | |
98% |
Safety and Handling
EINECSNumber : 244-536-5
TSCA : TSCA
RUO – Research Use Only