missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dibromoanisole, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 243230500

 View more versions of this product

Catalog No. AC243230500



Specifications

Specifications

2, 4-Dibromoanisole
Glass bottle
Beige
50g
21702-84-1
100.0
C7H6Br2O
MFCD00000079
XGXUGXPKRBQINS-UHFFFAOYSA-N
2,4-dibromo-1-methoxybenzene
27011
Crystalline Powder
Authentic
101.0°C to 106.0°C (1.0 mmHg)
60.0°C to 64.0°C
98%
97.5
98%
Br2(C6H3)OCH3
#, 2,4-dibromo-1-methoxy-benzene, 2,4-dibromoanisole, acmc-209fnf, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, chemwish ic06728, maybridge1_003667, pubchem3847, xgxugxpkrbqins-uhfffaoysa
COC1=C(C=C(C=C1)Br)Br
265.92
265.92
98%

Chemical Identifiers

21702-84-1
265.92
XGXUGXPKRBQINS-UHFFFAOYSA-N
27011
COC1=C(C=C(C=C1)Br)Br
C7H6Br2O
MFCD00000079
#, 2,4-dibromo-1-methoxy-benzene, 2,4-dibromoanisole, acmc-209fnf, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, chemwish ic06728, maybridge1_003667, pubchem3847, xgxugxpkrbqins-uhfffaoysa
2,4-dibromo-1-methoxybenzene
Safety and Handling

Safety and Handling

EINECSNumber : 244-536-5

TSCA : TSCA

SDS Documents