Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromoanisole, 98+%

$37.36 - $432.45

Chemical Identifiers

Chemical Name or Material 2,4-Dibromoanisole
CAS 21702-84-1
Molecular Formula C7H6Br2O
Formula Weight 265.93
MDL Number MFCD00000079
Beilstein 2045290
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1620014

 
alfa aesar™
A1620014
25g Each for $37.36

AAA1620022

 
alfa aesar™
A1620022
100g Each for $111.95

AAA1620036

 
alfa aesar™
A1620036
500g Each for $432.45
Description & Specifications

Specifications

Formula Weight 265.93
MDL Number MFCD00000079
Chemical Name or Material 2,4-Dibromoanisole
Melting Point 60° to 64°C
Beilstein 2045290
Quantity 25g
CAS 21702-84-1
Boiling Point 82° to 88°C (0.3mmHg)
Assay ≥98%
Molecular Formula C7H6Br2O