missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromoaniline, 98+%

Chemical Identifiers

CAS 615-57-6
Molecular Formula C6H5Br2N
Formula Weight 250.93
MDL Number MFCD00007633
UN Number UN2811
Beilstein 2206653
Chemical Name or Material 2,4-Dibromoaniline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1332906 SDS Alfa Aesar™
A1332906
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1332914 SDS Alfa Aesar™
A1332914
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1332922 SDS Alfa Aesar™
A1332922
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

615-57-6
250.93
UN2811
2,4-Dibromoaniline
C6H5Br2N
MFCD00007633
2206653

Specifications

615-57-6
156°C (24mmHg)
C6H5Br2N
250.93
UN2811
2,4-Dibromoaniline
78° to 82°C
5g
Characteristic
MFCD00007633
2206653
≥98%

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA