missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4'-Dibromoacetophenone, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC112682500
Specifications
2, 4'-Dibromoacetophenone | |
Glass bottle | |
Yellow to Beige | |
250g | |
97.5 | |
97.5% min. (GC) | |
BrC6H4COCH2Br | |
07, 285 | |
15, 1438 | |
Solubility in water: insoluble. Other solubilities: very soluble in warm alcohol,soluble in ethyl alcohol,ethyl ether,chloroform | |
C1=CC(=CC=C1C(=O)CBr)Br | |
277.94 | |
277.94 | |
98% |
Authentic | |
1415°C | |
107°C to 111°C | |
99-73-0 | |
100.0 | |
C8H6Br2O | |
MFCD00000200 | |
01,77; 03,34 | |
2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4'-bromophenacyl bromide, 4-bromo bromoacetyl benzene, 4-bromophenacyl bromide, alpha,4-dibromoacetophenone, alpha,p-dibromoacetophenone, ethanone, 2-bromo-1-4-bromophenyl, p-bromophenacyl bromide, p-bromophenacyl-8 | |
FKJSFKCZZIXQIP-UHFFFAOYSA-N | |
2-bromo-1-(4-bromophenyl)ethanone | |
7454 | |
Crystalline Solid |
Chemical Identifiers
99-73-0 | |
277.94 | |
FKJSFKCZZIXQIP-UHFFFAOYSA-N | |
7454 | |
C1=CC(=CC=C1C(=O)CBr)Br |
C8H6Br2O | |
MFCD00000200 | |
2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4'-bromophenacyl bromide, 4-bromo bromoacetyl benzene, 4-bromophenacyl bromide, alpha,4-dibromoacetophenone, alpha,p-dibromoacetophenone, ethanone, 2-bromo-1-4-bromophenyl, p-bromophenacyl bromide, p-bromophenacyl-8 | |
2-bromo-1-(4-bromophenyl)ethanone |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 202-783-6