missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromo-6-nitroaniline, 99%

$72.97 - $273.99

Chemical Identifiers

CAS 827-23-6
Molecular Formula C6H4Br2N2O2
Formula Weight 295.92
MDL Number MFCD00012107
Beilstein 2805574
Chemical Name or Material 2,4-Dibromo-6-nitroaniline
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2033306 SDS Alfa Aesar™
L2033306
5g Each for $72.97
AAL2033314 SDS Alfa Aesar™
L2033314
25g Each for $273.99
Specifications Description & Specifications

Chemical Identifiers

827-23-6
295.92
2805574
C6H4Br2N2O2
MFCD00012107
2,4-Dibromo-6-nitroaniline

Specifications

827-23-6
5g
295.92
2805574
99%
129° to 131°C
C6H4Br2N2O2
MFCD00012107
2,4-Dibromo-6-nitroaniline

Safety and Handling

GHS H Statement
H301-H311-H373-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger