Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromo-6-chloroaniline, 97%

$37.36 - $139.07

Chemical Identifiers

Chemical Name or Material 2,4-Dibromo-6-chloroaniline
CAS 874-18-0
Molecular Formula C6H4Br2ClN
Formula Weight 285.38
MDL Number MFCD00031419
Beilstein 2718195
Synonym 2-Chloro-4,6-dibromoaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1899806

  SDS
alfa aesar™
A1899806
5g Each for $37.36

AAA1899814

  SDS
alfa aesar™
A1899814
25g Each for $139.07
Description & Specifications

Specifications

Formula Weight 285.38
Melting Point 99° to 101°C
Molecular Formula C6H4Br2ClN
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Quantity 5g
MDL Number MFCD00031419
Assay 97%
Chemical Name or Material 2,4-Dibromo-6-chloroaniline
CAS 874-18-0
Beilstein 2718195
Synonym 2-Chloro-4,6-dibromoaniline
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.