Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dibromo-1-fluorobenzene, tech. 90%

$49.57 - $49.57

Chemical Identifiers

Chemical Name or Material 2,4-Dibromo-1-fluorobenzene
CAS 1435-53-6
Purity Grade Technical Grade
Molecular Formula C6H3Br2F
Formula Weight 253.91
MDL Number MFCD00000283
Beilstein 1934822
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2095506

  SDS
alfa aesar™
B2095506
5g Each for $49.57
Description & Specifications

Specifications

Formula Weight 253.91
Boiling Point 103°C (32mmHg)
Assay 90%
CAS 1435-53-6
Chemical Name or Material 2,4-Dibromo-1-fluorobenzene
Purity Grade Technical Grade
Quantity 5g
Flash Point 92°C (197°F)
Molecular Formula C6H3Br2F
MDL Number MFCD00000283
Beilstein 1934822
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.