Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazine, 97%

$157.00 - $157.00

Chemical Identifiers

Chemical Name or Material 2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazine
CAS 43167-79-9
Molecular Formula C19H20N6O
Formula Weight 348.41
MDL Number MFCD00272362
Synonym AAMT; 2,4-Bis(anilino)-6-(4-morpholinyl)-1,3,5-triazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2574203

 
alfa aesar™
B2574203
1g Each for $157.00
Description & Specifications

Specifications

Formula Weight 348.41
Chemical Name or Material 2,4-Dianilino-6-(4-morpholinyl)-1,3,5-triazine
Melting Point 194° to 196°C
Assay 97%
Quantity 1g
Molecular Formula C19H20N6O
MDL Number MFCD00272362
CAS 43167-79-9
Synonym AAMT; 2,4-Bis(anilino)-6-(4-morpholinyl)-1,3,5-triazine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.