missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Diaminothiazole hydrochloride, 95%

$54.74 - $714.00

Chemical Identifiers

CAS 72792-54-2
Molecular Formula C3H6ClN3S
Molecular Weight (g/mol) 151.612
MDL Number MFCD01438626
InChI Key PQAUAFCGXNRKIU-UHFFFAOYSA-N
Synonym thiazole-2,4-diamine hydrochloride, 1,3-thiazole-2,4-diamine hydrochloride, thiazole-2,4-diamine hcl, 2-thiazolamine, 4,5-dihydro-4-imino-, monohydrochloride, 4-thiazolamine, 2,5-dihydro-2-imino-, hydrochloride 1:1, 1,3-thiazole-2,4-diamine hydrochloride 1:1, 2,4-diaminothiazole, monohydrochloride, 1,3-thiazole-2,4-diaminehydrochloride, 1,3-thiazole-2,4-diamine hydrochloride 1:1
PubChem CID 118994
IUPAC Name 1,3-thiazole-2,4-diamine;hydrochloride
SMILES C1=C(N=C(S1)N)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65434MD View Documents Alfa Aesar™
H65434MD
250mg Each for $54.74
 
AAH6543403 View Documents Alfa Aesar™
H6543403
1g Each for $163.46
 
AAH6543406 View Documents Alfa Aesar™
H6543406
5g Each for $714.00
 
Specifications

Chemical Identifiers

72792-54-2
151.612
PQAUAFCGXNRKIU-UHFFFAOYSA-N
118994
C1=C(N=C(S1)N)N.Cl
C3H6ClN3S
MFCD01438626
thiazole-2,4-diamine hydrochloride, 1,3-thiazole-2,4-diamine hydrochloride, thiazole-2,4-diamine hcl, 2-thiazolamine, 4,5-dihydro-4-imino-, monohydrochloride, 4-thiazolamine, 2,5-dihydro-2-imino-, hydrochloride 1:1, 1,3-thiazole-2,4-diamine hydrochloride 1:1, 2,4-diaminothiazole, monohydrochloride, 1,3-thiazole-2,4-diaminehydrochloride, 1,3-thiazole-2,4-diamine hydrochloride 1:1
1,3-thiazole-2,4-diamine;hydrochloride

Specifications

250mg
C3H6ClN3S
3691448
PQAUAFCGXNRKIU-UHFFFAOYSA-N
1,3-thiazole-2,4-diamine;hydrochloride
118994
Solid
2,4-Diaminothiazole hydrochloride
72792-54-2
MFCD01438626
thiazole-2,4-diamine hydrochloride, 1,3-thiazole-2,4-diamine hydrochloride, thiazole-2,4-diamine hcl, 2-thiazolamine, 4,5-dihydro-4-imino-, monohydrochloride, 4-thiazolamine, 2,5-dihydro-2-imino-, hydrochloride 1:1, 1,3-thiazole-2,4-diamine hydrochloride 1:1, 2,4-diaminothiazole, monohydrochloride, 1,3-thiazole-2,4-diaminehydrochloride, 1,3-thiazole-2,4-diamine hydrochloride 1:1
C1=C(N=C(S1)N)N.Cl
151.612
151.61
95%