Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Diaminoquinazoline, 98+%

$48.34 - $143.78

Chemical Identifiers

Chemical Name or Material 2,4-Diaminoquinazoline
CAS 1899-48-5
Molecular Formula C8H8N4
Formula Weight 160.18
MDL Number MFCD00236032
Beilstein 131578
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1378303

 
alfa aesar™
L1378303
1g Each for $48.34

AAL1378306

 
alfa aesar™
L1378306
5g Each for $143.78
Description & Specifications

Specifications

Formula Weight 160.18
Chemical Name or Material 2,4-Diaminoquinazoline
CAS 1899-48-5
Molecular Formula C8H8N4
MDL Number MFCD00236032
Beilstein 131578
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Melting Point 254° to 258°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.