Learn More
2,4-Diaminophenol dihydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 212840250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 4-Diaminophenol dihydrochloride | |
97.5 | |
222°C | |
Authentic | |
Glass bottle | |
(H2N)2C6H3OH·2HCl | |
25 g | |
14, 2983 | |
Solubility in water: 27.5g/100mL (15°C). Other solubilities: slightly soluble in alcohol and ether | |
[H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1 | |
197.06 | |
197.07 | |
Crystalline Powder |
137-09-7 | |
100.0 | |
Beige to Gray-Green | |
97.5% min. (Argentometry) | |
C6H10Cl2N2O | |
MFCD00012979 | |
13, 550 | |
2,4-diaminophenol dihydrochloride, amidol, dianol, acrol, 2,4-diaminophenol hcl, 2,4-diaminophenol.2hcl, phenol, 2,4-diamino-, dihydrochloride, 2,4-diaminophenol hydrochloride, diamidophenol hydrochloride, unii-t1iyy101nx | |
KQEIJFWAXDQUPR-UHFFFAOYSA-N | |
2,4-diaminophenol;dihydrochloride | |
8715 | |
98% |
Chemical Identifiers
137-09-7 | |
197.06 | |
KQEIJFWAXDQUPR-UHFFFAOYSA-N | |
8715 | |
[H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1 |
C6H10Cl2N2O | |
MFCD00012979 | |
2,4-diaminophenol dihydrochloride, amidol, dianol, acrol, 2,4-diaminophenol hcl, 2,4-diaminophenol.2hcl, phenol, 2,4-diamino-, dihydrochloride, 2,4-diaminophenol hydrochloride, diamidophenol hydrochloride, unii-t1iyy101nx | |
2,4-diaminophenol;dihydrochloride |
Safety and Handling
GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica
GHS Signal Word: Danger
EINECSNumber : 205-279-4
RUO – Research Use Only