Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Diamino-6-undecyl-1,3,5-triazine, 97%

$62.68 - $910.08

Chemical Identifiers

Chemical Name or Material 2,4-Diamino-6-undecyl-1,3,5-triazine
CAS 2533-34-8
Molecular Formula C14H27N5
Formula Weight 265.4
MDL Number MFCD00213901
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1939203

  SDS
alfa aesar™
A1939203
1g Each for $62.68

AAA1939214

  SDS
alfa aesar™
A1939214
25g Each for $910.08
Description & Specifications

Specifications

Formula Weight 265.4
Chemical Name or Material 2,4-Diamino-6-undecyl-1,3,5-triazine
Assay 97%
Molecular Formula C14H27N5
MDL Number MFCD00213901
CAS 2533-34-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XY7070000
Melting Point 119° to 121°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.