Please login to your online account to display your discounted pricing

2,4-Diamino-6-phenyl-s-triazine ≥99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 4-Diamino-6-phenyl-s-triazine
Name Note 99+%
CAS 91-76-9
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H9N5
Formula Weight 187.2
MDL Number MFCD00023187
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC406405000

  SDS
thermo fisher scientific
406405000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 187.2
Quantity 500g
Color White
EINECS Number 202-095-6
TSCA TSCA
Density 1.4000g/mL
RTECS Number XY7000000
Melting Point 225° to 229°C
Beilstein 26, IV, 1244
Merck Index 15, 1091
CAS Min % 99.0
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 4-Diamino-6-phenyl-s-triazine
Packaging Glass bottle
Molecular Formula C9H9N5
CAS 91-76-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Infrared Spectrum Authentic
Name Note 99+%
Assay Percent Range 99+%
MDL Number MFCD00023187
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.