Please login to your online account to display your discounted pricing

2,4-Diamino-6-hydroxypyrimidine 96%, ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  112245000

 View more versions of this product

Catalog No. AC112245000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 126.12
Assay ≥95%
Physical Form Fine Crystalline Powder
Quantity 500g
Color White to cream
Beilstein 24, 469
Merck Index 15, 2983
TSCA TSCA
EINECS Number 200-254-4
CAS Min % 95.0
CAS Max % 100.0
Synonym 2, 6-Diamino-4-pyrimidinol
Infrared Spectrum Authentic
Water 0.5% max. (K.F.)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 96%
Name Note 96%
MDL Number MFCD00006098
CAS 56-06-4
Chemical Name or Material 2, 4-Diamino-6-hydroxypyrimidine
Molecular Formula C4H6N4O

Specifications

Chemical Name or Material 2, 4-Diamino-6-hydroxypyrimidine
Name Note 96%
CAS 56-06-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C4H6N4O
Formula Weight 126.12
MDL Number MFCD00006098
Synonym 2, 6-Diamino-4-pyrimidinol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents