Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Diamino-6-hydroxy-5-nitrosopyrimidine, 97%

$52.88 - $669.00

Chemical Identifiers

Chemical Name or Material 2,4-Diamino-6-hydroxy-5-nitrosopyrimidine
CAS 2387-48-6
Molecular Formula C4H5N5O2
Formula Weight 155.12
MDL Number MFCD00023263
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1014714

  SDS
alfa aesar™
A1014714
25g Each for $52.88

AAA1014722

  SDS
alfa aesar™
A1014722
100g Each for $180.81

AAA1014736

  SDS
alfa aesar™
A1014736
500g Each for $669.00
Description & Specifications

Specifications

Formula Weight 155.12
Chemical Name or Material 2,4-Diamino-6-hydroxy-5-nitrosopyrimidine
Assay 97%
Molecular Formula C4H5N5O2
MDL Number MFCD00023263
CAS 2387-48-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point >360°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.