missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Diamino-6-hydroxy-5-nitrosopyrimidine, 97%

Manufacturer:  Alfa Aesar™ A1014736

 View more versions of this product

Catalog No. AAA1014736



Specifications

Specifications

2,4-Diamino-6-hydroxy-5-nitrosopyrimidine
500g
C4H5N5O2
2,4-diamino-6-hydroxy-5-nitrosopyrimidine, 2,6-diamino-5-nitrosopyrimidin-4-ol, 4-pyrimidinol, 2,6-diamino-5-nitroso, 4 1h-pyrimidinone, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitroso-4-pyrimidinol, 4-pyrimidol, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitrosopyrimidin-4 3h-one, d04tlb, acmc-1cg71, 4-pyrimidol,6-diamino-5-nitroso
C1(=C(NC(=NC1=O)N)N)N=O
155.117
155.12
>360°C
2387-48-6
MFCD00023263
HVMRLFSFHWCUCG-UHFFFAOYSA-N
2,6-diamino-5-nitroso-1H-pyrimidin-4-one
75436
97%

Chemical Identifiers

2387-48-6
155.117
HVMRLFSFHWCUCG-UHFFFAOYSA-N
75436
C1(=C(NC(=NC1=O)N)N)N=O
C4H5N5O2
MFCD00023263
2,4-diamino-6-hydroxy-5-nitrosopyrimidine, 2,6-diamino-5-nitrosopyrimidin-4-ol, 4-pyrimidinol, 2,6-diamino-5-nitroso, 4 1h-pyrimidinone, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitroso-4-pyrimidinol, 4-pyrimidol, 2,6-diamino-5-nitroso, 2,6-diamino-5-nitrosopyrimidin-4 3h-one, d04tlb, acmc-1cg71, 4-pyrimidol,6-diamino-5-nitroso
2,6-diamino-5-nitroso-1H-pyrimidin-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents