missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Diamino-6-diallylamino-1,3,5-triazine 98.0+%, TCI America™
$154.71 - $1289.92
Chemical Identifiers
| CAS | 91-77-0 |
|---|---|
| Molecular Formula | C9H14N6 |
| Molecular Weight (g/mol) | 206.253 |
| MDL Number | MFCD00047347 |
| InChI Key | ROHTVIURAJBDES-UHFFFAOYSA-N |
| Synonym | N2,N2-Diallylmelamine |
| PubChem CID | 66682 |
| IUPAC Name | 2-N,2-N-bis(prop-2-enyl)-1,3,5-triazine-2,4,6-triamine |
| SMILES | C=CCN(CC=C)C1=NC(=NC(=N1)N)N |
Chemical Identifiers
| 91-77-0 | |
| 206.253 | |
| ROHTVIURAJBDES-UHFFFAOYSA-N | |
| 66682 | |
| C=CCN(CC=C)C1=NC(=NC(=N1)N)N |
| C9H14N6 | |
| MFCD00047347 | |
| N2,N2-Diallylmelamine | |
| 2-N,2-N-bis(prop-2-enyl)-1,3,5-triazine-2,4,6-triamine |
Specifications
| 91-77-0 | |
| White | |
| MFCD00047347 | |
| N2,N2-Diallylmelamine | |
| C=CCN(CC=C)C1=NC(=NC(=N1)N)N | |
| 206.253 | |
| 206.25 | |
| Crystalline Powder |
| 145°C | |
| C9H14N6 | |
| 25 g | |
| ROHTVIURAJBDES-UHFFFAOYSA-N | |
| 2-N,2-N-bis(prop-2-enyl)-1,3,5-triazine-2,4,6-triamine | |
| 66682 | |
| ≥98.0% (HPLC,T) | |
| 2,4-Diamino-6-diallylamino-1,3,5-triazine |
Safety and Handling
TSCA : Yes