Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Diamino-6-chloropyrimidine, 98%

$186.84 - $186.84

Chemical Identifiers

Chemical Name or Material 2,4-Diamino-6-chloropyrimidine
CAS 156-83-2
Molecular Formula C4H5ClN4
Formula Weight 144.56
MDL Number MFCD00006097
Beilstein 118455
Synonym 4-Chloro-2,6-pyrimidinediamine; 4-Chloro-2,6-diaminopyrimidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1778014

  SDS
alfa aesar™
A1778014
25g Each for $186.84

AAA1778006

  SDS
alfa aesar™
A1778006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2,4-Diamino-6-chloropyrimidine
Melting Point 199° to 202°C
Quantity 25g
CAS 156-83-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C4H5ClN4
Formula Weight 144.56
MDL Number MFCD00006097
Beilstein 118455
Synonym 4-Chloro-2,6-pyrimidinediamine; 4-Chloro-2,6-diaminopyrimidine
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.