missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Diamino-6-butylamino-1,3,5-triazine 97.0+%, TCI America™
$86.27 - $431.33
Chemical Identifiers
CAS | 5606-24-6 |
---|---|
Molecular Formula | C7H14N6 |
Molecular Weight (g/mol) | 182.23 |
MDL Number | MFCD00059764 |
InChI Key | CVKGSDYWCFQOKU-UHFFFAOYSA-N |
Synonym | 2-Butylamino-4,6-diamino-1,3,5-triazine, N2-Butylmelamine |
PubChem CID | 197831 |
IUPAC Name | N2-butyl-1,3,5-triazine-2,4,6-triamine |
SMILES | CCCCNC1=NC(N)=NC(N)=N1 |
Chemical Identifiers
5606-24-6 | |
182.23 | |
CVKGSDYWCFQOKU-UHFFFAOYSA-N | |
197831 | |
CCCCNC1=NC(N)=NC(N)=N1 |
C7H14N6 | |
MFCD00059764 | |
2-Butylamino-4,6-diamino-1,3,5-triazine, N2-Butylmelamine | |
N2-butyl-1,3,5-triazine-2,4,6-triamine |
Specifications
5606-24-6 | |
C7H14N6 | |
5 g | |
CVKGSDYWCFQOKU-UHFFFAOYSA-N | |
N2-butyl-1,3,5-triazine-2,4,6-triamine | |
197831 | |
≥97.0% (T) | |
2,4-Diamino-6-butylamino-1,3,5-triazine |
White-Yellow | |
MFCD00059764 | |
2-Butylamino-4,6-diamino-1,3,5-triazine, N2-Butylmelamine | |
CCCCNC1=NC(N)=NC(N)=N1 | |
182.23 | |
182.23 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : XY5852000
TSCA : No