Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Diamino-5-fluoroquinazoline, 97%

$59.38 - $230.54

Chemical Identifiers

Chemical Name or Material 2,4-Diamino-5-fluoroquinazoline
CAS 119584-70-2
Molecular Formula C8H7FN4
Formula Weight 178.17
MDL Number MFCD00221785
Beilstein 3590571
Synonym 5-Fluoroquinazoline-2,4-diamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2088003

  SDS
alfa aesar™
B2088003
1g Each for $59.38

AAB2088006

  SDS
alfa aesar™
B2088006
5g Each for $230.54
Description & Specifications

Specifications

Formula Weight 178.17
Synonym 5-Fluoroquinazoline-2,4-diamine
RTECS Number VA1367730
MDL Number MFCD00221785
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Chemical Name or Material 2,4-Diamino-5-fluoroquinazoline
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 119584-70-2
Quantity 1g
Health Hazard 1 Warning
Melting Point 245° to 248°C
Beilstein 3590571
Molecular Formula C8H7FN4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.