Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,4-Di(tert-pentyl)phenoxyacetic acid, 98%

$50.18 - $717.31

Chemical Identifiers

Chemical Name or Material 2,4-Di(tert-pentyl)phenoxyacetic acid
CAS 13402-96-5
Molecular Formula C18H28O3
Formula Weight 292.42
MDL Number MFCD00128799
Synonym 2,4-Di(tert-amyl)phenoxyacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1854614

 
alfa aesar™
A1854614
25g Each for $50.18

AAA1854622

 
alfa aesar™
A1854622
100g Each for $152.39

AAA1854636

 
alfa aesar™
A1854636
500g Each for $717.31
Description & Specifications

Specifications

Formula Weight 292.42
Chemical Name or Material 2,4-Di(tert-pentyl)phenoxyacetic acid
TSCA TSCA
Assay 98%
Molecular Formula C18H28O3
MDL Number MFCD00128799
CAS 13402-96-5
Synonym 2,4-Di(tert-amyl)phenoxyacetic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 125° to 127°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.