Please login to your online account to display your discounted pricing

2,4-D (For EPA 500, 8000/Appendix IX Methods)


2,4-D at 100μg/mL in methanol

Quantity 1x1mL (in Methanol)
Packaging Ampule

Manufacturer: ultra scientific  HB1001

 View more versions of this product

Catalog No. US-HB-100-1

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1x1mL (in Methanol)
Alpha Vector D
Methods For EPA 500, 8000/ Appendix IX Methods
Packaging Ampule
CAS 94-75-7
Chemical Name or Material 2, 4-D

Specifications

Chemical Name or Material 2, 4-D
CAS 94-75-7