missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-D (For EPA 500, 8000/Appendix IX Methods)

2,4-D at 100μg/mL in methanol

$46.40 - $63.06

Chemical Identifiers

CAS 94-75-7
Chemical Name or Material 2, 4-D
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-HB-100-1 Ultra Scientific
HB1001
Ampule 1x1mL (in Methanol) Each for $46.40
US-PST-140 Ultra Scientific
PST140
Clear or Amber Vial 100mg (Neat) Each for $63.06
Specifications Description & Specifications

Chemical Identifiers

94-75-7
2, 4-D

Specifications

1x1mL (in Methanol)
For EPA 500, 8000/ Appendix IX Methods
94-75-7
D
Ampule
2, 4-D