Please login to your online account to display your discounted pricing

2,4-D (For EPA 500, 8000/Appendix IX Methods)


2,4-D at 100μg/mL in methanol

Quantity 100mg (Neat)
Packaging Clear or Amber Vial

Manufacturer: ultra scientific  PST140

 View more versions of this product

Catalog No. US-PST-140

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100mg (Neat)
Methods For EPA 500, 8000/ Appendix IX Methods
Alpha Vector D
Chemical Name or Material 2, 4-D
CAS 94-75-7
Packaging Clear or Amber Vial

Specifications

Chemical Name or Material 2, 4-D
CAS 94-75-7